1,7-Diazaspiro[4.5]decane-7-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 1,7-diazaspiro[4.5]decane-7-carboxylate

070566-250MG 070566-500MG 070566-1G
101945-358EA 1221.54 USD
101945-358 101945-360 101945-362
1,7-Diazaspiro[4.5]decane-7-carboxylic acid tert-butyl ester ≥95%
1,7-Diazaspiro[4.5]decane-7-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₃H₂₄N₂O₂
MW: 240.35 g/mol
MDL Number: MFCD08752633
CAS Number: 939793-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR