2,5-Diazaspiro[3.5]nonane-5-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

070565-100MG 070565-250MG 070565-500MG
101945-352EA 573.85 USD
101945-352 101945-354 101945-356
2,5-Diazaspiro[3.5]nonane-5-carboxylic acid tert-butyl ester ≥95%
2,5-Diazaspiro[3.5]nonane-5-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₂
MDL Number: MFCD14581199
CAS Number: 1246035-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR