2-Aminomethyl-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Aminomethyl-7-Boc-7-azaspiro[3.5]nonane

070564-100MG 070564-250MG 070564-500MG
101945-346EA 524.62 USD
101945-346 101945-348 101945-350
2-Aminomethyl-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester ≥95%
2-Aminomethyl-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₄H₂₆N₂O₂
MW: 254.38 g/mol
MDL Number: MFCD12198554
CAS Number: 1160247-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR