tert-Butyl 2,8-diazaspiro[4.5]decane-8-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2,8-Diazaspiro[4.5]decane-8-carboxylic acid tert-butyl ester hydrochloride, 8-Boc-2,8-Diazaspiro[4.5]decane hydrochloride

070562-250MG 070562-1G
101945-336EA 155.38 USD
101945-336 102833-482
tert-Butyl 2,8-diazaspiro[4.5]decane-8-carboxylate hydrochloride ≥95%
tert-Butyl 2,8-diazaspiro[4.5]decane-8-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₅ClN₂O₂
MW: 276.81 g/mol
MDL Number: MFCD13185062
CAS Number: 851325-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR