1,8-Diazaspiro[4.5]decane-8-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 1,8-diazaspiro[4.5]decane-8-carboxylate

070561-100MG 070561-250MG
101945-328EA 524.62 USD
101945-328 101945-330
1,8-Diazaspiro[4.5]decane-8-carboxylic acid tert-butyl ester ≥95%
1,8-Diazaspiro[4.5]decane-8-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₃H₂₄N₂O₂
MW: 240.35 g/mol
MDL Number: MFCD11501187
CAS Number: 937729-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR