4-Oxohexahydrocyclopenta[c]pyrrole-2-carboxylic acid tert-butyl ester ≥96%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 4-oxohexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate, Hexahydrocyclopenta[c]pyrrol-4(1H)-one, N-BOC protected
070559-250MG
070559-500MG
070559-1G
101945-316EA
815.38
USD
101945-316
101945-318
101945-320
4-Oxohexahydrocyclopenta[c]pyrrole-2-carboxylic acid tert-butyl ester ≥96%
4-Oxohexahydrocyclopenta[c]pyrrole-2-carboxylic acid tert-butyl ester
Hazard: Irritant
Formula:
C₁₂H₁₉NO₃ MW: 225.29 g/mol |
MDL Number:
MFCD06658358 CAS Number: 879686-42-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...