1,6-Diazaspiro[3.5]nonane-6-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 1,6-diazaspiro[3.5]nonane-6-carboxylate

070556-100MG 070556-250MG 070556-500MG
101945-298EA 446.15 USD
101945-298 101945-300 101945-302
1,6-Diazaspiro[3.5]nonane-6-carboxylic acid tert-butyl ester ≥95%
1,6-Diazaspiro[3.5]nonane-6-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₂
MW: 226.32 g/mol
MDL Number: MFCD14581197
CAS Number: 1251002-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR