3-Oxo-1-oxa-7-azaspiro[4.4]nonane-7-carboxylic acid tert-butyl ester ≥96%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-oxo-1-oxa-7-azaspiro[4.4]nonane-7-carboxylate

070555-250MG 070555-500MG 070555-1G
101945-292EA 1047.69 USD
101945-292 101945-294 101945-296
3-Oxo-1-oxa-7-azaspiro[4.4]nonane-7-carboxylic acid tert-butyl ester ≥96%
3-Oxo-1-oxa-7-azaspiro[4.4]nonane-7-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₂H₁₉NO₄
MW: 241.29 g/mol
MDL Number: MFCD12198523
CAS Number: 1160246-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR