1,7-Diazaspiro[4.4]nonane-1-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

070554-100MG 070554-250MG 070554-500MG
101945-286EA 524.62 USD
101945-286 101945-288 101945-290
1,7-Diazaspiro[4.4]nonane-1-carboxylic acid tert-butyl ester ≥95%
1,7-Diazaspiro[4.4]nonane-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₂
MDL Number: MFCD08234442
CAS Number: 885268-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR