2-Azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2-azaspiro[3.3]heptane-2-carboxylate

070549-100MG 070549-250MG 070549-500MG
101945-256EA 466.15 USD
101945-256 101945-258 101945-260
2-Azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester ≥95%
2-Azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₁H₁₉NO₂
MW: 197.28 g/mol
MDL Number: MFCD18325169
CAS Number: 1259489-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR