4-Bromo-2-methoxypyrimidine ≥98%

Supplier: Matrix Scientific

070546-1G 070546-5G
101945-242EA 287.69 USD
101945-242 101945-244
4-Bromo-2-methoxypyrimidine ≥98%
4-Bromo-2-methoxypyrimidine

Hazard: Irritant

Formula: C₅H₅BrN₂O
MW: 189.0100 g/mol
MDL Number: MFCD09879097
CAS Number: 959240-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR