1-tert-Butylsulfinyl-5-oxa-1-azaspiro[2.3]hexane ≥95%

Supplier: Matrix Scientific

070545-250MG 070545-500MG 070545-1G
101945-236EA 1221.54 USD
101945-236 101945-238 101945-240
1-tert-Butylsulfinyl-5-oxa-1-azaspiro[2.3]hexane ≥95%
1-tert-Butylsulfinyl-5-oxa-1-azaspiro[2.3]hexane

Hazard: Irritant

Formula: C₈H₁₅NO₂S
MDL Number: MFCD18325167
CAS Number: 1211284-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR