3,6-Dihydro-4-(tributylstannyl)-1(2H)-pyridinecarboxylic acid tert-butyl ester ≥94%

Supplier: Matrix Scientific

070542-1G 070542-5G
101945-226EA 1047.69 USD
101945-226 101945-228
3,6-Dihydro-4-(tributylstannyl)-1(2H)-pyridinecarboxylic acid tert-butyl ester ≥94%
3,6-Dihydro-4-(tributylstannyl)-1(2H)-pyridinecarboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₆H₃₀O₂Sn
MDL Number: MFCD18325165
CAS Number: 208465-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR