3-Bromo-1-methyl-1H-pyrazole ≥95%
Supplier: Matrix Scientific
070535-1G
070535-5G
101945-198EA
44.62
USD
101945-198
101945-200
3-Bromo-1-methyl-1H-pyrazole ≥95%
3-Bromo-1-methyl-1H-pyrazole
Hazard: Irritant/Refrigerate
Formula:
C4H5BrN2 MW: 161.0000 g/mol |
MDL Number:
MFCD11110668 CAS Number: 151049-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...