3-Bromo-6-chloroimidazo[1,2-b]pyridazine

Supplier: Matrix Scientific

070532-5G 070532-25G
101945-186EA 60 USD
101945-186 101945-188
3-Bromo-6-chloroimidazo[1,2-b]pyridazine
3-Bromo-6-chloroimidazo[1,2-b]pyridazine

Hazard: Irritant

Formula: C₆H₃BrClN₃
MDL Number: MFCD09027274
CAS Number: 13526-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR