6-Chloro-3-iodoimidazo[1,2-a]pyridazine ≥96%

Supplier: Matrix Scientific

070531-1G 070531-5G
101945-180EA 1396.92 USD
101945-180 101945-182
6-Chloro-3-iodoimidazo[1,2-a]pyridazine ≥96%
6-Chloro-3-iodoimidazo[1,2-a]pyridazine

Hazard: Irritant

Formula: C₆H₃ClIN₃
MDL Number: MFCD11044749
CAS Number: 923595-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR