6-chloro-3-iodoimidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Chloro-3-iodoimidazo[1,2-a]pyridine

070529-1G 070529-5G
101945-172EA 1221.54 USD
101945-172 101945-174
6-chloro-3-iodoimidazo[1,2-a]pyridine ≥95%
6-chloro-3-iodoimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₄ClIN₂
MW: 278.477 g/mol
MDL Number: MFCD06739217
CAS Number: 885275-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR