Dihydro-2H-thiopyran-3(4H)-one-1,1-dioxide ≥95%

Supplier: Matrix Scientific

070523-1G 070523-5G
101945-146EA 1221.54 USD
101945-146 101945-148
Dihydro-2H-thiopyran-3(4H)-one-1,1-dioxide ≥95%
Dihydro-2H-thiopyran-3(4H)-one-1,1-dioxide

Hazard: Irritant

Formula: C₅H₈O₃S
MDL Number: MFCD16622215
CAS Number: 29431-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR