4,4-Divinyl-p-biphenyl
Supplier: Matrix Scientific
Synonyms:
4,4'-Divinyl-1,1'-biphenyl
070522-1G
070522-5G
101945-142EA
1396.92
USD
101945-142
101945-144
4,4-Divinyl-p-biphenyl
4,4-Divinyl-p-biphenyl
Hazard: Irritant
Formula:
C₁₆H₁₄ MW: 206.29 g/mol Boiling Pt: 341 °C Density: 0.998 Flash Pt: 170 °C |
MDL Number:
MFCD18325139 CAS Number: 4433-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...