(1H-Indazol-5-yl)methanol

Supplier: Matrix Scientific

Synonyms: 5-(Hydroxymethyl)-1H-indazole

070521-1G 070521-5G
101945-138EA 307.69 USD
101945-138 101945-140
(1H-Indazol-5-yl)methanol
(1H-Indazol-5-yl)methanol

Hazard: Irritant

Formula: C₈H₈N₂O
MW: 148.1600 g/mol
MDL Number: MFCD04038636
CAS Number: 478828-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR