Azetidin-3-ylmethanol ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Hydroxymethyl)azetidine

070519-100MG 070519-250MG 070519-500MG
101945-128EA 223.62 USD
101945-128 101945-130 101945-132
Azetidin-3-ylmethanol ≥97%
Azetidin-3-ylmethanol

Hazard: Irritant

Formula: C₄H₉NO
MW: 87.12 g/mol
MDL Number: MFCD08703352
CAS Number: 95849-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR