7-(Tributylstannyl)[1,2,4]triazolo[1,5-a]pyridine ≥95%

Supplier: Matrix Scientific

070518-1G 070518-5G
101945-124EA 1803.08 USD
101945-124 101945-126
7-(Tributylstannyl)[1,2,4]triazolo[1,5-a]pyridine ≥95%
7-(Tributylstannyl)[1,2,4]triazolo[1,5-a]pyridine

Hazard: Irritant

Formula: C₁₈H₃₁N₃Sn
MDL Number: MFCD17015011
CAS Number: 1245816-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR