8-(Tributylstannyl)[1,2,4]triazolo[1,5-a]pyridine ≥96%

Supplier: Matrix Scientific

070517-1G 070517-5G
101945-120EA 1300 USD
101945-120 101945-122
8-(Tributylstannyl)[1,2,4]triazolo[1,5-a]pyridine ≥96%
8-(Tributylstannyl)[1,2,4]triazolo[1,5-a]pyridine

Hazard: Irritant

Formula: C₁₈H₃₁N₃Sn
CAS Number: 1245816-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR