4-Fluoro-7-azaindole ≥96%

Supplier: Matrix Scientific

070516-1G 070516-5G
101945-116EA 376.92 USD
101945-116 101945-118
4-Fluoro-7-azaindole ≥96%
4-Fluoro-7-azaindole

Hazard: Irritant

Formula: C₇H₅FN₂
MDL Number: MFCD08272231
CAS Number: 640735-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR