7-(Tributylstannyl)isoquinoline ≥94%

Supplier: Matrix Scientific

070513-1G 070513-5G
101945-108EA 1038.46 USD
101945-108 101945-110
7-(Tributylstannyl)isoquinoline ≥94%
7-(Tributylstannyl)isoquinoline

Hazard: Irritant

Formula: C₂₁H₃₃NSn
MDL Number: MFCD17015017
CAS Number: 1161975-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR