8-(Tributylstannyl)isoquinoline ≥94%
Supplier: Matrix Scientific
070512-1G
070512-5G
101945-104EA
1221.54
USD
101945-104
101945-106
8-(Tributylstannyl)isoquinoline ≥94%
8-(Tributylstannyl)isoquinoline
Hazard: Irritant
Formula:
C₂₁H₃₃NSn |
MDL Number:
MFCD17015016 CAS Number: 1245816-24-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...