5-(Tributylstannyl)isoquinoline ≥94%

Supplier: Matrix Scientific

070511-1G 070511-5G
101945-100EA 2561.54 USD
101945-100 101945-102
5-(Tributylstannyl)isoquinoline ≥94%
5-(Tributylstannyl)isoquinoline

Hazard: Irritant

Formula: C₂₁H₃₃NSn
MDL Number: MFCD17015013
CAS Number: 1161976-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR