2-Chloropyrimidine-4-carboxylic acid ≥95%
Supplier: Matrix Scientific
070507-1G
070507-5G
101945-084EA
75.38
USD
101945-084
101945-086
2-Chloropyrimidine-4-carboxylic acid ≥95%
2-Chloropyrimidine-4-carboxylic acid
Hazard: Irritant
Formula:
C₅H₃ClN₂O₂ MW: 158.5400 g/mol |
MDL Number:
MFCD06739041 CAS Number: 149849-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...