3-Phenyl-2-(phenylsulfonyl)-1,2-oxaziridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Benzenesulphonyl)-3-phenyloxaziridine

070506-1G 070506-5G
101945-080EA 187.69 USD
101945-080 101945-082
3-Phenyl-2-(phenylsulfonyl)-1,2-oxaziridine ≥95%
3-Phenyl-2-(phenylsulfonyl)-1,2-oxaziridine

Hazard: Irritant/Refrigerate

Formula: C13H11NO3S
CAS Number: 63160-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR