3-Iodo-1H-pyrazolo[3,4-b]pyridine ≥96%

Supplier: Matrix Scientific

Synonyms: 3-Iodo1H-pyrazolo[3,4-b]pyridine

070505-1G 070505-5G
101945-076EA 75.38 USD
101945-076 101945-078
3-Iodo-1H-pyrazolo[3,4-b]pyridine ≥96%
3-Iodo-1H-pyrazolo[3,4-b]pyridine

Hazard: Irritant

Formula: C₆H₄IN₃
MW: 245.02 g/mol
MDL Number: MFCD09749818
CAS Number: 117007-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR