4-Bromo-2-fluoro-3-iodopyridine

Supplier: Matrix Scientific

070504-1G 070504-5G
101945-072EA 930.77 USD
101945-072 101945-074
4-Bromo-2-fluoro-3-iodopyridine
4-Bromo-2-fluoro-3-iodopyridine

Hazard: Irritant

Formula: C₅H₂BrFIN
MDL Number: MFCD17014987
CAS Number: 917969-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR