4-Chloro-1H-pyrazolo[3,4-b]pyridine

Supplier: Matrix Scientific

070503-500MG 070503-1G
101945-068EA 1221.54 USD
101945-068 101945-070
4-Chloro-1H-pyrazolo[3,4-b]pyridine
4-Chloro-1H-pyrazolo[3,4-b]pyridine

Hazard: Irritant

Formula: C₆H₄ClN₃
MDL Number: MFCD11846313
CAS Number: 29274-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR