4-Chloro-6-iodothieno[3,2-d]pyrimidine ≥95%

Supplier: Matrix Scientific

070502-1G 070502-5G
101945-064EA 1221.54 USD
101945-064 101945-066
4-Chloro-6-iodothieno[3,2-d]pyrimidine ≥95%
4-Chloro-6-iodothieno[3,2-d]pyrimidine

Hazard: Irritant

Formula: C₆H₂ClIN₂S
MDL Number: MFCD09746329
CAS Number: 225382-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR