7-Bromo-2-methyl-2H-indazole-3-carboxaldehyde

Supplier: Matrix Scientific

Synonyms: 7-Bromo-2-methyl-2H-indazole-3-carbaldehyde

070499-1G 070499-5G
101945-052EA 1241.54 USD
101945-052 101945-054
7-Bromo-2-methyl-2H-indazole-3-carboxaldehyde
7-Bromo-2-methyl-2H-indazole-3-carboxaldehyde

Hazard: Irritant

Formula: C₉H₇BrN₂O
MW: 239.08 g/mol
MDL Number: MFCD11520881
CAS Number: 845751-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR