5-Bromo-α,α-dimethyl-2-thiazolemethanol ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-a,a-dimethyl-2-thiazolemethanol

070497-1G 070497-5G
101945-044EA 930.77 USD
101945-044 101945-046
5-Bromo-α,α-dimethyl-2-thiazolemethanol ≥97%
5-Bromo-alpha,alpha-dimethyl-2-thiazolemethanol

Hazard: Irritant

Formula: C₆H₈BrNOS
MW: 222.11 g/mol
MDL Number: MFCD18325135
CAS Number: 879488-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR