α,α-Dimethyl-2-thiazolemethanol ≥94%
Supplier: Matrix Scientific
Synonyms:
2-Thiazol-2-yl-propan-2-ol
070496-1G
070496-5G
101945-040EA
815.38
USD
101945-040
101945-042
α,α-Dimethyl-2-thiazolemethanol ≥94%
alpha,alpha-Dimethyl-2-thiazolemethanol
Hazard: Irritant
Formula:
C₆H₉NOS MW: 143.21 g/mol |
MDL Number:
MFCD03211903 CAS Number: 16077-78-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...