(S)-tert-Butyl-4,4-difluoro-2-(hydroxymethyl)pyrrolidine-1-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: (S)-tert-Butyl 4,4-difluoro-2-(hydroxymethyl)pyrrolidine-1-carboxylate

070495-1G 070495-5G
101945-036EA 1592.31 USD
101945-036 101945-038
(S)-tert-Butyl-4,4-difluoro-2-(hydroxymethyl)pyrrolidine-1-carboxylate ≥97%
(S)-tert-Butyl-4,4-difluoro-2-(hydroxymethyl)pyrrolidine-1-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₇F₂NO₃
MW: 237.25 g/mol
MDL Number: MFCD12755205
CAS Number: 215918-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR