4-Chloro-6-(tributylstannyl)thieno[3,2-d]pyrimidine ≥90%

Supplier: Matrix Scientific

070494-1G 070494-5G
101945-032EA 815.38 USD
101945-032 101945-034
4-Chloro-6-(tributylstannyl)thieno[3,2-d]pyrimidine ≥90%
4-Chloro-6-(tributylstannyl)thieno[3,2-d]pyrimidine

Hazard: Irritant

Formula: C₁₈H₂₉ClN₂SSn
MDL Number: MFCD18325134
CAS Number: 875339-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR