5-(Aminomethyl)-1H-indazole 97

Supplier: Matrix Scientific

070493-1G 070493-5G
101945-028EA 815.38 USD
101945-028 101945-030
5-(Aminomethyl)-1H-indazole 97
5-(Aminomethyl)-1H-indazole

Hazard: Irritant

Formula: C₈H₉N₃
MW: 147.18 g/mol
MDL Number: MFCD09028157
CAS Number: 267413-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR