3-(Aminomethyl)-5-fluoropyridine ≥96%
Supplier: Matrix Scientific
Synonyms:
3-Aminomethyl-5-fluoropyridine
070492-1G
070492-5G
101945-024EA
1338.46
USD
101945-024
101945-026
3-(Aminomethyl)-5-fluoropyridine ≥96%
3-(Aminomethyl)-5-fluoropyridine
Hazard: Irritant
Formula:
C₆H₇FN₂ MW: 126.130 g/mol |
MDL Number:
MFCD08450424 CAS Number: 23586-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...