4-Methoxy-3,6-dihydro-2H-pyran ≥95%

Supplier: Matrix Scientific

Synonyms: 5,6-Dihydro-4-methoxy-2H-pyran

070491-1G 070491-5G
101945-020EA 176.92 USD
101945-020 101945-022
4-Methoxy-3,6-dihydro-2H-pyran ≥95%
4-Methoxy-3,6-dihydro-2H-pyran

Hazard: Irritant

Formula: C₆H₁₀O₂
MW: 114.15 g/mol
MDL Number: MFCD00006570
CAS Number: 17327-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR