2,5-Bis(tributylstannyl)furan

Supplier: Matrix Scientific

070489-1G 070489-5G
101945-012EA 584.62 USD
101945-012 101945-014
2,5-Bis(tributylstannyl)furan
2,5-Bis(tributylstannyl)furan

Hazard: Irritant

Formula: C₂₈H₅₆OSn₂
MDL Number: MFCD09833740
CAS Number: 193361-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR