2-Acetylamino-5-thiazolecarboxylic acid methyl ester ≥97%

Supplier: Matrix Scientific

070488-1G 070488-5G
101945-008EA 349.23 USD
101945-008 101945-010
2-Acetylamino-5-thiazolecarboxylic acid methyl ester ≥97%
2-Acetylamino-5-thiazolecarboxylic acid methyl ester

Hazard: Irritant

Formula: C₇H₈N₂O₃S
MDL Number: MFCD17015003
CAS Number: 1174534-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR