5-Bromo-2-(tert-butyldimethylsilyl)-4-methylthiazole

Supplier: Matrix Scientific

070487-1G 070487-5G
101945-004EA 510.77 USD
101945-004 101945-006
5-Bromo-2-(tert-butyldimethylsilyl)-4-methylthiazole
5-Bromo-2-(tert-butyldimethylsilyl)-4-methylthiazole

Hazard: Irritant

Formula: C₁₀H₁₈BrNSSi
MW: 292.32 g/mol
MDL Number: MFCD17015004
CAS Number: 1245782-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR