Thieno[3,2-b]pyridin-3-amine ≥96%

Supplier: Matrix Scientific

Synonyms: 3-Aminothieno[3,2-b]pyridine

070485-1G 070485-5G
101944-996EA 815.38 USD
101944-996 101944-998
Thieno[3,2-b]pyridin-3-amine ≥96%
Thieno[3,2-b]pyridin-3-amine

Hazard: Irritant

Formula: C₇H₆N₂S
MW: 150.2 g/mol
MDL Number: MFCD11109671
CAS Number: 120208-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR