4-Bromoisoquinolin-1(2H)-one ≥96%

Supplier: Matrix Scientific

070483-1G 070483-5G
101944-988EA 815.38 USD
101944-988 101944-990
4-Bromoisoquinolin-1(2H)-one ≥96%
4-Bromoisoquinolin-1(2H)-one

Hazard: Irritant

Formula: C₉H₆BrNO
MDL Number: MFCD08437036
CAS Number: 3951-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR