5-Bromo-2-chloro-4-(tributylstannyl)pyridine

Supplier: Matrix Scientific

070478-1G 070478-5G
101944-968EA 640 USD
101944-968 101944-970
5-Bromo-2-chloro-4-(tributylstannyl)pyridine
5-Bromo-2-chloro-4-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₇H₂₉BrClNSn
MDL Number: MFCD17015005
CAS Number: 821773-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR