1-Boc-6-fluoro-1H-indole ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Boc-6-Fluoro-1H-indole

070475-1G 070475-5G
101944-960EA 640 USD
101944-960 101944-962
1-Boc-6-fluoro-1H-indole ≥97%
1-Boc-6-fluoro-1H-indole

Hazard: Irritant

Formula: C₁₃H₁₄FNO₂
MW: 235.2542 g/mol
MDL Number: MFCD09909713
CAS Number: 1208459-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR