Benzothiazole-5-carboxylic acid ≥96%

Supplier: Matrix Scientific

070472-1G 070472-5G
101944-948EA 476.92 USD
101944-948 101944-950
Benzothiazole-5-carboxylic acid ≥96%
Benzothiazole-5-carboxylic acid

Hazard: Irritant

Formula: C₈H₅NO₂S
MDL Number: MFCD06796380
CAS Number: 68867-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR