4-Methyl-2H-pyridazin-3-one ≥99%

Supplier: Matrix Scientific

Synonyms: 4-Methylpyridazin-3(2H)-one

070471-1G 070471-5G 070471-25G
101944-942EA 150.77 USD
101944-942 101944-944 101944-946
4-Methyl-2H-pyridazin-3-one ≥99%
4-Methyl-2H-pyridazin-3-one

Hazard: Irritant

Formula: C₅H₆N₂O
MW: 110.12 g/mol
MDL Number: MFCD08460985
CAS Number: 33471-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR